Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hmha1
Entrez Gene ID
314618
Gene Name
histocompatibility (minor) HA-1
Gene Aliases
RGD1308662
Chromosome Location
Chr.7: 12541056 - 12541812 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108067.1 NP_001101537.1 - 2467 122

View Full Product Details