Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arid3a
Entrez Gene ID
314616
Gene Name
AT rich interactive domain 3A (Bright like)
Gene Aliases
-
Chromosome Location
Chr.7: 12575989 - 12602415 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108066.2 NP_001101536.2 7 - 8 1815 58
XM_006240902.2 XP_006240964.1 7 - 8 2130 58
XM_006240903.1 XP_006240965.1 7 - 8 1948 58
XM_006240904.2 XP_006240966.1 7 - 8 1950 58
XM_006240905.2 XP_006240967.1 7 - 8 2313 58
XM_008765169.1 XP_008763391.1 7 - 8 1821 58
GenBank mRNA BC168846.1 - 7 - 8 1815 58

View Full Product Details