Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Asgr1
Entrez Gene ID
24210
Gene Name
asialoglycoprotein receptor 1
Gene Aliases
ASGR, RATRHL1, RHL1
Chromosome Location
Chr.10: 56662191 - 56666086 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012503.2 NP_036635.1 6 - 7 503 98
XM_006246579.1 XP_006246641.1 6 - 7 615 98
XM_008767768.1 XP_008765990.1 6 - 7 635 98
XM_008767769.1 XP_008765991.1 6 - 7 579 98
GenBank mRNA BC088154.1 - 6 - 7 503 98
FQ209763.1 - 6 - 7 539 98
FQ218553.1 - 6 - 7 579 98
FQ218644.1 - 6 - 7 539 98
FQ219107.1 - 6 - 7 576 98
FQ219710.1 - 6 - 7 620 98
M21770.1 - 5 - 6 405 98

View Full Product Details