Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stk32b
Entrez Gene ID
305431
Gene Name
serine/threonine kinase 32B
Gene Aliases
-
Chromosome Location
Chr.14: 77837073 - 78097584 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107224.2 NP_001100694.1 - 663 63

View Full Product Details