Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gas2l1
Entrez Gene ID
360973
Gene Name
growth arrest-specific 2 like 1
Gene Aliases
-
Chromosome Location
Chr.14: 85308443 - 85318487 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108365.1 NP_001101835.1 2 - 3 346 106

View Full Product Details