The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_054011.1 | NP_446463.2 | 8 - 9 | 1159 | 89 |
XM_008770976.1 | XP_008769198.1 | 6 - 7 | 3021 | 89 | |
GenBank mRNA | AB027562.1 | - | 8 - 9 | 1159 | 89 |
FQ210473.1 | - | 9 - 10 | 1377 | 89 |