Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Papolb
Entrez Gene ID
304300
Gene Name
poly (A) polymerase beta (testis specific)
Gene Aliases
-
Chromosome Location
Chr.12: 14112426 - 14114772 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012020.2 NP_001012020.1 1 - 1 2249 69
GenBank mRNA BC079046.1 - 1 - 1 2249 69
CR458765.1 - 1 - 1 69

View Full Product Details