Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf6
Entrez Gene ID
304271
Gene Name
ring finger protein (C3H2C3 type) 6
Gene Aliases
-
Chromosome Location
Chr.12: 10574168 - 10583196 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107118.1 NP_001100588.1 5 - 6 677 95

View Full Product Details