Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc22a18
Entrez Gene ID
309131
Gene Name
solute carrier family 22, member 18
Gene Aliases
-
Chromosome Location
Chr.1: 216676331 - 216700632 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004260.1 NP_001004260.1 2 - 3 291 104
XM_006230775.2 XP_006230837.1 2 - 3 350 104
GenBank mRNA BC079159.1 - 2 - 3 291 104
FQ228759.1 - 2 - 3 320 104
FQ231035.1 - 2 - 3 320 104

View Full Product Details