Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pip5k1b
Entrez Gene ID
309419
Gene Name
phosphatidylinositol-4-phosphate 5-kinase, type I, beta
Gene Aliases
Pip5k1a
Chromosome Location
Chr.1: 242168462 - 242441247 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012743.1 NP_001012761.1 9 - 10 1582 81
XM_008760304.1 XP_008758526.1 9 - 10 1213 81
GenBank mRNA BC090349.1 - 9 - 10 1582 81

View Full Product Details