Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Uevld
Entrez Gene ID
691172
Gene Name
UEV and lactate/malate dehyrogenase domains
Gene Aliases
-
Chromosome Location
Chr.1: 102982833 - 103014480 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008759461.1 XP_008757683.1 8 - 9 1028 56
XM_008759462.1 XP_008757684.1 8 - 9 1027 56

View Full Product Details