Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bnc2
Entrez Gene ID
298189
Gene Name
basonuclin 2
Gene Aliases
-
Chromosome Location
Chr.5: 102414000 - 102743417 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106666.1 NP_001100136.1 2 - 3 219 64
XM_008763775.1 XP_008761997.1 1 - 2 1410 64
XM_008763776.1 XP_008761998.1 1 - 2 1410 64
XM_008763777.1 XP_008761999.1 1 - 2 1409 64

View Full Product Details