The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024797.1 | NP_001019968.1 | 32 - 33 | 6885 | 88 |
XM_006232735.2 | XP_006232797.1 | 32 - 33 | 6876 | 88 | |
XM_006232736.2 | XP_006232798.1 | 32 - 33 | 6861 | 88 | |
XM_006232737.2 | XP_006232799.1 | 31 - 32 | 6840 | 88 | |
XM_006232738.2 | XP_006232800.1 | 31 - 32 | 6837 | 88 | |
XM_006232739.2 | XP_006232801.1 | 32 - 33 | 6816 | 88 | |
XM_006232740.2 | XP_006232802.1 | 31 - 32 | 6672 | 88 | |
XM_006232741.2 | XP_006232803.1 | 31 - 32 | 6666 | 88 | |
XM_008761201.1 | XP_008759423.1 | 33 - 34 | 6890 | 88 | |
XM_008761202.1 | XP_008759424.1 | 31 - 32 | 6325 | 88 | |
GenBank mRNA | AB195689.1 | - | 31 - 32 | 6719 | 88 |
AY724781.1 | - | 32 - 33 | 6885 | 88 | |
AY724782.1 | - | 31 - 32 | 6965 | 88 |