The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106440.1 | NP_001099910.1 | 2 - 3 | 495 | 94 |
XM_006232609.2 | XP_006232671.1 | 3 - 4 | 437 | 94 | |
GenBank mRNA | BC158876.1 | - | 3 - 4 | 402 | 94 |