The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100976.1 | NP_001094446.1 | 9 - 10 | 2113 | 70 |
XM_006232668.1 | XP_006232730.1 | 10 - 11 | 2240 | 70 | |
XM_006232669.1 | XP_006232731.1 | 10 - 11 | 2219 | 70 | |
XM_008761174.1 | XP_008759396.1 | 10 - 11 | 2264 | 70 | |
XM_008761175.1 | XP_008759397.1 | 9 - 10 | 2135 | 70 | |
GenBank mRNA | BC085776.1 | - | 8 - 9 | 3002 | 70 |
BC129100.1 | - | 9 - 10 | 2113 | 70 |