Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dap3
Entrez Gene ID
295238
Gene Name
death associated protein 3
Gene Aliases
-
Chromosome Location
Chr.2: 188225607 - 188255327 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011950.2 NP_001011950.2 5 - 6 680 64
XM_006232610.2 XP_006232672.1 5 - 6 476 64
XM_006232611.2 XP_006232673.1 5 - 6 476 64
GenBank mRNA BC085878.1 - 5 - 6 680 64

View Full Product Details