Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbp
Entrez Gene ID
117526
Gene Name
TATA box binding protein
Gene Aliases
TFIID
Chromosome Location
Chr.1: 57491385 - 57509335 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004198.1 NP_001004198.1 7 - 8 1145 156
XM_006227980.2 XP_006228042.1 7 - 8 1088 156
GenBank mRNA BC081939.1 - 7 - 8 1145 156

View Full Product Details