Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zc3hav1
Entrez Gene ID
252832
Gene Name
zinc finger CCCH type, antiviral 1
Gene Aliases
ARTD13, Zap
Chromosome Location
Chr.4: 66012111 - 66062156 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006236295.2 XP_006236357.1 9 - 10 2700 60

View Full Product Details