Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbxas1
Entrez Gene ID
24886
Gene Name
thromboxane A synthase 1, platelet
Gene Aliases
-
Chromosome Location
Chr.4: 66624181 - 66846745 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012687.1 NP_036819.1 2 - 3 314 94
XM_008762759.1 XP_008760981.1 2 - 3 375 94
XM_008762761.1 XP_008760983.1 2 - 3 365 94
GenBank mRNA D28773.1 - 2 - 3 314 94
D31798.1 - 2 - 3 236 94

View Full Product Details