The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034117.1 | NP_001029289.1 | 11 - 12 | 1374 | 97 |
NM_053739.2 | NP_446191.1 | 11 - 12 | 1396 | 97 | |
GenBank mRNA | AY033824.1 | - | 10 - 11 | 1172 | 97 |
BC074011.1 | - | 11 - 12 | 1261 | 97 | |
FQ230174.1 | - | 2 - 3 | 614 | 97 |