Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stx8
Entrez Gene ID
59074
Gene Name
syntaxin 8
Gene Aliases
-
Chromosome Location
Chr.10: 54512278 - 54753302 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031656.2 NP_113844.1 7 - 8 664 63
XM_008767863.1 XP_008766085.1 8 - 9 888 63
GenBank mRNA AF033109.1 - 7 - 8 669 63

View Full Product Details