Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plk4
Entrez Gene ID
310344
Gene Name
polo-like kinase 4
Gene Aliases
-
Chromosome Location
Chr.2: 127686912 - 127705518 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107669.1 NP_001101139.1 3 - 4 405 106

View Full Product Details