Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nosip
Entrez Gene ID
292894
Gene Name
nitric oxide synthase interacting protein
Gene Aliases
-
Chromosome Location
Chr.1: 101055571 - 101072938 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006229025.1 XP_006229087.1 1 - 2 98 60
GenBank mRNA FQ216351.1 - 1 - 2 65 60

View Full Product Details