Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ambp
Entrez Gene ID
25377
Gene Name
alpha-1-microglobulin/bikunin precursor
Gene Aliases
-
Chromosome Location
Chr.5: 78975690 - 78986020 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012901.1 NP_037033.1 7 - 8 101
XM_008763759.1 XP_008761981.1 7 - 8 794 101
GenBank mRNA BC088166.1 - 7 - 8 693 101
FQ209788.1 - 4 - 5 276 101
FQ209831.1 - 4 - 5 276 101
FQ210037.1 - 4 - 5 276 101
FQ210364.1 - 4 - 5 101
FQ219366.1 - 4 - 5 276 101
FQ219628.1 - 4 - 5 277 101
FQ219767.1 - 4 - 5 276 101
S87544.1 - 7 - 8 101

View Full Product Details