Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Donson
Entrez Gene ID
288257
Gene Name
downstream neighbor of SON
Gene Aliases
-
Chromosome Location
Chr.11: 31834644 - 31847490 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008287.1 NP_001008288.1 1 - 2 413 66
XM_006248047.2 XP_006248109.1 1 - 2 407 66
XM_006248048.2 XP_006248110.1 1 - 2 407 66
GenBank mRNA BC085847.1 - 1 - 2 413 66

View Full Product Details