The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130579.1 | NP_001124051.1 | 11 - 12 | 933 | 69 |
XM_008765775.1 | XP_008763997.1 | 11 - 12 | 919 | 69 |