Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
A1bg
Entrez Gene ID
140656
Gene Name
alpha-1-B glycoprotein
Gene Aliases
C44
Chromosome Location
Chr.7: 102294805 - 102298522 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022258.2 NP_071594.2 7 - 8 1565 141
GenBank mRNA AJ302031.1 - 7 - 8 1565 141
BC089771.1 - 7 - 8 1548 141

View Full Product Details