Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prcc
Entrez Gene ID
310687
Gene Name
papillary renal cell carcinoma (translocation-associated)
Gene Aliases
-
Chromosome Location
Chr.2: 187232458 - 187258140 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107700.1 NP_001101170.1 6 - 7 1562 78
GenBank mRNA BC166708.1 - 6 - 7 1562 78

View Full Product Details