Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ttc24
Entrez Gene ID
100361582
Gene Name
tetratricopeptide repeat domain 24
Gene Aliases
-
Chromosome Location
Chr.2: 187430557 - 187438471 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006232788.2 XP_006232850.1 10 - 11 1749 77
XM_006232789.2 XP_006232851.1 10 - 11 1746 77
XM_008761231.1 XP_008759453.1 9 - 10 1669 77

View Full Product Details