The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107680.1 | NP_001101150.1 | 7 - 8 | 816 | 92 |
XM_006232477.2 | XP_006232539.1 | 6 - 7 | 779 | 92 | |
XM_006232478.2 | XP_006232540.1 | - | 717 | 92 | |
XM_006232479.2 | XP_006232541.1 | 5 - 6 | 695 | 92 | |
GenBank mRNA | FQ215154.1 | - | 7 - 8 | 819 | 92 |
FQ215514.1 | - | 7 - 8 | 813 | 92 | |
FQ216377.1 | - | 7 - 8 | 777 | 92 | |
FQ216411.1 | - | 7 - 8 | 845 | 92 | |
FQ224275.1 | - | 4 - 5 | 474 | 92 |