Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il5
Entrez Gene ID
24497
Gene Name
interleukin 5
Gene Aliases
-
Chromosome Location
Chr.10: 39066716 - 39069587 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021834.1 NP_068606.1 1 - 2 132 145
GenBank mRNA AJ011299.1 - 1 - 2 132 145

View Full Product Details