Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Naglu
Entrez Gene ID
360630
Gene Name
N-acetylglucosaminidase, alpha
Gene Aliases
RGD1564228
Chromosome Location
Chr.10: 88979365 - 88986879 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002727815.5 XP_002727861.2 1 - 2 434 109

View Full Product Details