Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dhx58
Entrez Gene ID
303538
Gene Name
DEXH (Asp-Glu-X-His) box polypeptide 58
Gene Aliases
Lgp2, RGD1310093
Chromosome Location
Chr.10: 88599722 - 88611565 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098788.1 NP_001092258.1 6 - 7 991 129
XM_006247314.2 XP_006247376.1 7 - 8 1560 129
XM_006247315.2 XP_006247377.1 7 - 8 1549 129

View Full Product Details