Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abca17
Entrez Gene ID
287112
Gene Name
ATP-binding cassette, subfamily A (ABC1), member 17
Gene Aliases
-
Chromosome Location
Chr.10: 13630076 - 13723214 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031637.1 NP_001026807.1 20 - 21 3441 76
XM_006245927.2 XP_006245989.1 21 - 22 4549 76
XM_006245928.2 XP_006245990.1 21 - 22 4533 76
XM_008767554.1 XP_008765776.1 21 - 22 5163 76
XM_008767556.1 XP_008765778.1 20 - 21 3422 76
GenBank mRNA AB196699.1 - 20 - 21 3441 76

View Full Product Details