Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rhbdd3
Entrez Gene ID
289753
Gene Name
rhomboid domain containing 3
Gene Aliases
RGD1311827
Chromosome Location
Chr.14: 85350983 - 85357029 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006251204.2 XP_006251266.1 1 - 2 217 160

View Full Product Details