Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rhbdd3
Entrez Gene ID
289753
Gene Name
rhomboid domain containing 3
Gene Aliases
RGD1311827
Chromosome Location
Chr.14: 85350983 - 85357029 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013875.1 NP_001013897.1 6 - 7 1242 66
XM_006251204.2 XP_006251266.1 7 - 8 1339 66
XM_006251205.2 XP_006251267.1 6 - 7 1129 66
XM_006251206.2 XP_006251268.1 7 - 8 1172 66
XM_006251207.2 XP_006251269.1 6 - 7 1130 66
XM_006251208.2 XP_006251270.1 7 - 8 1279 66
XM_006251209.2 XP_006251271.1 6 - 7 1322 66
XM_006251210.2 XP_006251272.1 5 - 6 1118 66
GenBank mRNA BC081912.1 - 6 - 7 1242 66

View Full Product Details