The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107217.1 | NP_001100687.1 | 3 - 4 | 387 | 81 |
GenBank mRNA | FQ211206.1 | - | 3 - 4 | 349 | 81 |
FQ221610.1 | - | 3 - 4 | 351 | 81 |