The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019292.4 | NP_062165.2 | 6 - 7 | 678 | 70 |
XM_006232116.1 | XP_006232178.1 | 7 - 8 | 1003 | 70 | |
GenBank mRNA | AB030829.1 | - | 6 - 7 | 678 | 70 |
AB030830.1 | - | 6 - 7 | 678 | 70 | |
AF037072.1 | - | 6 - 7 | 701 | 70 | |
BC061980.1 | - | 6 - 7 | 713 | 70 | |
FQ210528.1 | - | 6 - 7 | 726 | 70 | |
FQ211459.1 | - | 6 - 7 | 725 | 70 | |
FQ214846.1 | - | 6 - 7 | 723 | 70 | |
FQ214929.1 | - | 6 - 7 | 728 | 70 | |
FQ215392.1 | - | 5 - 6 | 540 | 70 |