Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lgals7
Entrez Gene ID
29518
Gene Name
lectin, galactoside-binding, soluble, 7
Gene Aliases
-
Chromosome Location
Chr.1: 87043926 - 87047223 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006228670.2 XP_006228732.1 1 - 2 393 135

View Full Product Details