Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rasip1
Entrez Gene ID
292912
Gene Name
Ras interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 101608995 - 101621648 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106261.2 NP_001099731.1 2 - 3 396 61
XM_006229030.2 XP_006229092.1 4 - 5 1481 61
GenBank mRNA BC168876.1 - 2 - 3 396 61

View Full Product Details