The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024868.1 | NP_001020039.1 | 2 - 3 | 488 | 71 |
XM_006229027.1 | XP_006229089.1 | 3 - 4 | 225 | 71 | |
XM_006229028.2 | XP_006229090.1 | 3 - 4 | 461 | 71 | |
GenBank mRNA | BC097946.1 | - | 2 - 3 | 488 | 71 |