The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100752.1 | NP_001094222.1 | 10 - 11 | 970 | 60 |
XM_006237500.1 | XP_006237562.1 | 10 - 11 | 1053 | 60 | |
GenBank mRNA | BC091192.1 | - | 10 - 11 | 936 | 60 |
BC168777.1 | - | 10 - 11 | 970 | 60 | |
FQ216233.1 | - | 10 - 11 | 970 | 60 | |
FQ216671.1 | - | 10 - 11 | 960 | 60 | |
FQ216764.1 | - | 10 - 11 | 960 | 60 | |
FQ232722.1 | - | 10 - 11 | 968 | 60 | |
FQ233877.1 | - | 10 - 11 | 1039 | 60 |