The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100752.1 | NP_001094222.1 | 9 - 10 | 910 | 67 |
XM_006237500.1 | XP_006237562.1 | 9 - 10 | 993 | 67 | |
GenBank mRNA | BC091192.1 | - | 9 - 10 | 876 | 67 |
BC168777.1 | - | 9 - 10 | 910 | 67 | |
FQ216540.1 | - | 9 - 10 | 911 | 67 | |
FQ216671.1 | - | 9 - 10 | 900 | 67 | |
FQ216764.1 | - | 9 - 10 | 900 | 67 | |
FQ229931.1 | - | 1 - 2 | 136 | 67 | |
FQ232722.1 | - | 9 - 10 | 908 | 67 | |
FQ233877.1 | - | 9 - 10 | 979 | 67 |