Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slco1a6
Entrez Gene ID
84608
Gene Name
solute carrier organic anion transporter family, member 1a6
Gene Aliases
Oatp5, Slc21a13
Chromosome Location
Chr.4: 176345361 - 176390724 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130736.1 NP_570092.1 2 - 3 235 124
XM_006237650.2 XP_006237712.1 3 - 4 289 124
GenBank mRNA AF053317.2 - 2 - 3 235 124

View Full Product Details