The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014057.1 | NP_001014079.1 | 2 - 3 | 222 | 112 |
XM_008763240.1 | XP_008761462.1 | 2 - 3 | 245 | 112 | |
XM_008763241.1 | XP_008761463.1 | 2 - 3 | 284 | 112 | |
XM_008763242.1 | XP_008761464.1 | 2 - 3 | 246 | 112 | |
GenBank mRNA | BC087631.1 | - | 2 - 3 | 222 | 112 |