Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddx47
Entrez Gene ID
297685
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 47
Gene Aliases
-
Chromosome Location
Chr.4: 168772558 - 168787594 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015005.2 NP_001015005.2 1 - 2 100 81
XM_006237518.1 XP_006237580.1 2 - 3 157 81
GenBank mRNA BC091696.1 - 1 - 2 93 81
CB761683.1 - 1 - 2 112 81

View Full Product Details