Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aebp2
Entrez Gene ID
297705
Gene Name
AE binding protein 2
Gene Aliases
-
Chromosome Location
Chr.4: 174799383 - 174863260 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106626.1 NP_001100096.1 4 - 5 621 134
XM_006237595.2 XP_006237657.1 6 - 7 1337 134
XM_008763376.1 XP_008761598.1 6 - 7 1317 134
XM_008763377.1 XP_008761599.1 6 - 7 1338 134

View Full Product Details