The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108280.1 | NP_001101750.1 | 7 - 8 | 897 | 87 |
XM_006246751.1 | XP_006246813.1 | 6 - 7 | 927 | 87 | |
XM_006246752.2 | XP_006246814.1 | 6 - 7 | 1011 | 87 | |
XM_006246754.2 | XP_006246816.1 | 6 - 7 | 718 | 87 | |
XM_008767844.1 | XP_008766066.1 | 5 - 6 | 1081 | 87 |