Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Serpinf2
Entrez Gene ID
287527
Gene Name
serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 2
Gene Aliases
-
Chromosome Location
Chr.10: 62264247 - 62272476 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011892.1 NP_001011892.1 2 - 3 111 60
XM_006246675.1 XP_006246737.1 2 - 3 256 60
GenBank mRNA BC079362.1 - 2 - 3 111 60

View Full Product Details