Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrrc46
Entrez Gene ID
287653
Gene Name
leucine rich repeat containing 46
Gene Aliases
-
Chromosome Location
Chr.10: 84971674 - 84983785 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004201.1 NP_001004201.1 6 - 7 680 57
XM_006247268.2 XP_006247330.1 6 - 7 510 57
GenBank mRNA BC079258.1 - 6 - 7 680 57

View Full Product Details